Cobalt(II) sulfamate hydrate
Cobalt(II) sulfamate hydrate
Cobalt(II) sulfamate hydrate
Thermo Scientific Chemicals

Cobalt(II) sulfamate hydrate

CAS: 14017-41-5 | CoH4N2O6S2 | 251.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 043906.36
also known as 043906-36
Price (USD)
430.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
430.00
Each
Chemical Identifiers
CAS14017-41-5
IUPAC Nameλ²-cobalt(2+) disulfamate
Molecular FormulaCoH4N2O6S2
InChI KeyWLQXLCXXAPYDIU-UHFFFAOYSA-L
SMILES[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Red
Assay (special)18-20% [Cobalt (Co)]
Identification (FTIR)Conforms
Waters of hydration3-4%
Cobalt(II) sulfamate hydrate is used in photonic device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only