Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min
Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min
Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min
Thermo Scientific Chemicals

Potassium tetrachloroplatinate(II), Premion™, 99.99% (metals basis), Pt 46.4% min

CAS: 10025-99-7 | Cl4H4K2Pt | 419.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043946.06
also known as 043946-06
Price (USD)
785.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
785.00
Each
Chemical Identifiers
CAS10025-99-7
IUPAC Namedipotassium tetrachloroplatinumdiuide
Molecular FormulaCl4H4K2Pt
InChI KeyJCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[K+].[K+].[Pt--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max., Pt 46.4% min
Potassium tetrachloroplatinate(II) is used as important reagent for the preparation of other coordination complexes of platinum. It is also used in the induction of apoptosis in human breast cancer cells. Also used in the preparation of camptothecin-linked platinum anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only