Silver Protein for Histology, Strong
Thermo Scientific Chemicals

Silver Protein for Histology, Strong

CAS: 9008-42-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
5g
25g
110g
Catalog number 043970.06
also known as 043970-06
Price (USD)
-
Quantity:
5g
Request bulk or custom format
Chemical Identifiers
CAS9008-42-8
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Silver content: 7.5-8.5%
Appearance (Color)Pale yellow to brown
FormPowder
AssaySilver content: 7.5-8.5%
Silver protein is used in electron microscopy with periodic acid and thiocarbohydrazide or thiosemicarbohydrazide as a positive stain for carbohydrates such as glycogen. It can also be used for light microscopy to stain nerve tissue. It is also used in histological stain for microscopic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only