Germanium pieces, 3.2mm (0.125in) and down, 99.999% (metals basis)
Germanium pieces, 3.2mm (0.125in) and down, 99.999% (metals basis)
Germanium pieces, 3.2mm (0.125in) and down, 99.999% (metals basis)
Thermo Scientific Chemicals

Germanium pieces, 3.2mm (0.125in) and down, 99.999% (metals basis)

CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 043986.09
also known as 043986-09
Price (USD)
198.65
Special offer
Online exclusive
Ends: 28-Dec-2026
234.00
Save 35.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 28-Dec-2026
234.00
Save 35.35
Each
Chemical Identifiers
CAS7440-56-4
IUPAC Namegermanium
Molecular FormulaGe
InChI KeyGNPVGFCGXDBREM-UHFFFAOYSA-N
SMILES[Ge]
View more
SpecificationsSpecification SheetSpecification Sheet
Certificate of ConformanceCertificate of Conformance Provided
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only