Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%
Thermo Scientific Chemicals

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%

Vapor deposition | CAS: 14877-41-9 | C33H57CoO6 | 608.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 043993.09
also known as 043993-09
Price (USD)
684.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
684.00
Each
Specifications
CAS14877-41-9
Chemical Name or MaterialTris (2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt (III)
ColorGreen
Formula Weight608.72
Health Hazard 1H317-H351
View more
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III) is used precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only