Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis)
Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis)
Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Catalog NumberQuantity
043996.8V
also known as 043996-8V
60 m
Catalog number 043996.8V
also known as 043996-8V
Price (USD)
305.65
Online Exclusive
339.00
Save 33.35 (10%)
Each
Quantity:
60 m
Request bulk or custom format
Price (USD)
305.65
Online Exclusive
339.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
FormWire, 0.375mm (0.015in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only