Copper turnings, 99.9% (metals basis)
Copper turnings, 99.9% (metals basis)
Copper turnings, 99.9% (metals basis)
Thermo Scientific Chemicals

Copper turnings, 99.9% (metals basis)

Typically 1-12mm wide | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 044001.18
also known as 044001-18
Price (USD)
49.65
Online Exclusive
54.70
Save 5.05 (9%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.70
Save 5.05 (9%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormTurnings, 1-12mm wide.
Appearance (Color)Brown
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only