Aluminum oxide desiccant, regular
Aluminum oxide desiccant, regular
Aluminum oxide desiccant, regular
Thermo Scientific Chemicals

Aluminum oxide desiccant, regular

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 044018.22
also known as 044018-22
Price (USD)
37.20
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
37.20
Each
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment3.2mm Spheres
Aluminum oxide is used in dessicators ceramics, coating material and electrical insulators. It is an ingredient in sunscreen, sometimes present in cosmetics such as blush, lipstick, and nail polish. Aluminum oxide is the catalyst in the Claus process in hydro desulfurization and some Ziegler-Natta polymerizations. It is widely used as an abrasive, in the automotive industries, cosmetic industries and fiber materials.It is a medium for chromatography in some sodium vapor lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Dry gases and liquids
  • Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2
  • Liquids include: benzene, hexane, oil, pentane
RUO – Research Use Only