Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)
Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)
Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)
Thermo Scientific Chemicals

Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)

Form: Approx. 1.04g/mm; Cast rod, diameter is approximate | CAS: 7440-27-9 | Tb | 158.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
25 mm
Catalog number 044049.DH
also known as 044049-DH
Price (USD)
2,125.65
Special offer
Online exclusive
Ends: 30-Jun-2026
2,501.00
Save 375.35 (15%)
Each
Quantity:
25 mm
Request bulk or custom format
Price (USD)
2,125.65
Special offer
Online exclusive
Ends: 30-Jun-2026
2,501.00
Save 375.35 (15%)
Each
Chemical Identifiers
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max. excluding Ta
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only