Ammonium magnesium phosphate hydrate, 99.999% (metals basis)
Ammonium magnesium phosphate hydrate, 99.999% (metals basis)
Ammonium magnesium phosphate hydrate, 99.999% (metals basis)
Thermo Scientific Chemicals

Ammonium magnesium phosphate hydrate, 99.999% (metals basis)

CAS: 7785-21-9 | H4MgNO4P | 137.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 044053.30
also known as 044053-30
Price (USD)
507.65
Special offer
Online exclusive
Ends: 30-Jun-2026
597.00
Save 89.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
507.65
Special offer
Online exclusive
Ends: 30-Jun-2026
597.00
Save 89.35 (15%)
Each
Chemical Identifiers
CAS7785-21-9
IUPAC Namemagnesium(2+) ammonium phosphate
Molecular FormulaH4MgNO4P
InChI KeyMXZRMHIULZDAKC-UHFFFAOYSA-L
SMILES[NH4+].[Mg++].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Ammonium magnesium phosphate hydrate general application in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only