Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water
Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water
Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water
Thermo Scientific Chemicals

Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water

CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044062.18
Price (USD)
216.65
Special offer
Online exclusive
Ends: 30-Jun-2026
255.00
Save 38.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
216.65
Special offer
Online exclusive
Ends: 30-Jun-2026
255.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS2567-83-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥97.5% (dry weight basis)
Assay from Supplier's CofAWater content: ≥10.0 to ≤12.0%
Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only