Silver conductive adhesive paste
Thermo Scientific Chemicals

Silver conductive adhesive paste

 
Have Questions?
Catalog NumberQuantity
044075.18
also known as 044075-18
50 g
Catalog number 044075.18
also known as 044075-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver conductive adhesive paste
ColorSilver
EINECS Number232-033-3
Health Hazard 1H315-H319-H335-H336
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only