Bismuth(III) telluride, 99.98% (metals basis)
Bismuth(III) telluride, 99.98% (metals basis)
Bismuth(III) telluride, 99.98% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) telluride, 99.98% (metals basis)

CAS: 1304-82-1 | Bi2Te3
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 044077.06
also known as 044077-06
Price (USD)
72.65
Online Exclusive
80.50
Save 7.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.50
Save 7.85 (10%)
Each
Chemical Identifiers
CAS1304-82-1
IUPAC Namedibismuth(3+) tritellanediide
Molecular FormulaBi2Te3
InChI KeyAZFMNKUWQAGOBM-UHFFFAOYSA-N
SMILES[Te--].[Te--].[Te--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max.
Bismuth(III) telluride and its alloys (antimony or selenium) posses excellent semiconductor activity, which is used in thermoelectric industry for cooling and energy generation. It increases the speed of microchips and acts as topological insulators in spintronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only