Vanadium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)
Vanadium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)
Vanadium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Vanadium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 044105.G1
also known as 044105-G1
Price (USD)
85.60
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
85.60
Each
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Comment0.25mm diameter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only