Vanadium wire, 0.075mm (0.003in) dia, 99.8% (metals basis)
Vanadium wire, 0.075mm (0.003in) dia, 99.8% (metals basis)
Vanadium wire, 0.075mm (0.003in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Vanadium wire, 0.075mm (0.003in) dia, 99.8% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 044108.G1
also known as 044108-G1
Price (USD)
237.65
Online Exclusive
264.00
Save 26.35 (10%)
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
237.65
Online Exclusive
264.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silver
Assay from Supplier's CofA≥99.8% (metals basis)
FormWire, 0.075mm (0.003in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only