Cadmium foil, 0.05mm (0.002in) thick, Puratronic™, 99.999% (metals basis)
Cadmium foil, 0.05mm (0.002in) thick, Puratronic™, 99.999% (metals basis)
Cadmium foil, 0.05mm (0.002in) thick, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium foil, 0.05mm (0.002in) thick, Puratronic™, 99.999% (metals basis)

CAS: 7440-43-9 | Cd | 112.41 g/mol
Have Questions?
Catalog NumberQuantity
044127.FI
also known as 044127-FI
50 x 50 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044127.FI
also known as 044127-FI
Price (USD)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium
Molecular FormulaCd
InChI KeyBDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.05mm (0.002in) thick.
Appearance (Color)White silvery
Assay from Supplier's CofA≥99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only