Lithium methoxide, 98+%
Lithium methoxide, 98+%
Lithium methoxide, 98+%
Thermo Scientific Chemicals

Lithium methoxide, 98+%

CAS: 865-34-9 | CH3LiO | 37.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044129.36
also known as 044129-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS865-34-9
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% min
Lithium methoxide is used in sol-gel synthesis of lithium borophosphates. It is also useful for deprotonation reactions in organic synthesis. Further, it serves as a mild base in organic synthesis especially in transesterifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only