Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%
Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%
Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%
Thermo Scientific Chemicals

Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%

CAS: 17524-05-9 | C10H14MoO6 | 326.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 044140.09
also known as 044140-09
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.40
Save 7.75 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.40
Save 7.75 (15%)
Each
Chemical Identifiers
CAS17524-05-9
IUPAC Namedioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14MoO6
InChI KeySKMUJBBRXZPAJY-VGKOASNMSA-L
SMILESO=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder or crystals or crystalline powder
Assay from Suppliers CofAMolybdenum (Mo): 29.0-30.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Melting Point180.0-184.0°C (U.S. specification)
Forms a functional model compound for the study of oxotransferase molybdenum enzymes. Mild catalyst for the deprotection of acetals in high yield. Catalyzes high yield oxidation of secondary alcohols to ketones with sodium percarbonate and phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only