Methyl Red sodium salt, 0.02% w/v aq. soln.
Methyl Red sodium salt, 0.02% w/v aq. soln.
Methyl Red sodium salt, 0.02% w/v aq. soln.
Thermo Scientific Chemicals

Methyl Red sodium salt, 0.02% w/v aq. soln.

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 044161.AP
also known as 044161-AP
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.00
Save 8.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.00
Save 8.35 (15%)
Each
Specifications
Assay Percent Range0.02% w/v
CAS845-10-3
Chemical Name or MaterialMethyl Red sodium salt
EINECS Number212-682-9
Formula Weight291.29
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only