Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis) - FAQs
Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis) - FAQs
View additional product information for Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis) - FAQs (044164.FI, 044164.FF)
2 product FAQs found
What is the weight of the 50x50 mm sheet of Gold foil, 0.5mm thick, Premion, 99.999% (metals basis)?
The 50x50 mm sheet of gold foil weighs ~24 g.
What is the weight of the 25x25 mm sheet of Gold foil, 0.5mm thick, Premion, 99.999% (metals basis)?
The 25x25 mm sheet of gold foil weighs ~6 g.