Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis)
Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis)
Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals

Gold foil, 0.5mm (0.02in) thick, Premion™, 99.999% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 044164.FI
also known as 044164-FI
Price (USD)
5,597.00
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
5,597.00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold foil
EINECS Number231-165-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the weight of the 50x50 mm sheet of Gold foil, 0.5mm thick, Premion, 99.999% (metals basis)?

The 50x50 mm sheet of gold foil weighs ~24 g.

What is the weight of the 25x25 mm sheet of Gold foil, 0.5mm thick, Premion, 99.999% (metals basis)?

The 25x25 mm sheet of gold foil weighs ~6 g.