Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals

Moisture Sensitive; Tap density 1.5g/cm3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044173.A1
also known as 044173-A1
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities2.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the tap density of Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis)?

The tap density of Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis) is 1.4.

What is the iron wt% maximum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis)?

The iron wt% maximum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis) is 0.2?

What is the oil and grease wt% maximum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis)?

The oil and grease wt% maximum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis) is 0.2.

What is the volatile wt% maximum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis)?

The volatile wt% maximum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis) is 0.1.

What is the free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis)?

The free metallic aluminum wt% minimum for Aluminum powder, spherical, APS 4.5-7 micron, 97.5% (metals basis) is 97.5.