Tris(dibenzoylmethane)mono(phenanthroline)europium(III)
Tris(dibenzoylmethane)mono(phenanthroline)europium(III)
Tris(dibenzoylmethane)mono(phenanthroline)europium(III)
Thermo Scientific Chemicals

Tris(dibenzoylmethane)mono(phenanthroline)europium(III)

As an organic red light emitter | CAS: 17904-83-5 | C57H44EuN2O6 | 1004.951 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number 044175.02
also known as 044175-02
Price (USD)
364.65
Online Exclusive
405.00
Save 40.35 (10%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
364.65
Online Exclusive
405.00
Save 40.35 (10%)
Each
Chemical Identifiers
CAS17904-83-5
IUPAC Name1,10-phenanthroline tris(3-hydroxy-1,3-diphenylprop-2-en-1-one) europium
Molecular FormulaC57H44EuN2O6
InChI KeyDYKOLWWJTALFFU-UHFFFAOYSA-N
SMILES[Eu].C1=CN=C2C(C=CC3=CC=CN=C23)=C1.OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormPowder
Assay from Suppliers CofA≥98.5%
Melting Point183-192°C
As an organic red light emitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only