Zinc bromide hydrate, 99.9% (metals basis)
Thermo Scientific Chemicals

Zinc bromide hydrate, 99.9% (metals basis)

CAS: 299465-28-4 | Br2Zn | 225.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 044230.36
also known as 044230-36
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS299465-28-4
IUPAC Namezinc(2+) dibromide
Molecular FormulaBr2Zn
InChI KeyVNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES[Zn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Zinc bromide is an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. Zinc bromide is employed as a lewis acid in organic chemistry. Has been used as a transparent shield against radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only