Bismuth Lead eutectic wire, 0.79mm (0.031in) dia
Thermo Scientific Chemicals

Bismuth Lead eutectic wire, 0.79mm (0.031in) dia

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 lb.
1 lb.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044238.Q4
also known as 044238-Q4
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35 (10%)
Each
Quantity:
0.25 lb.
Request bulk or custom format
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35 (10%)
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeBi:Pb; 55.5:44.5 wt%
Chemical Name or MaterialBismuth Lead eutectic wire
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Employed in chemical research. LBE is suggested as a coolant in some nuclear reactors and is a proposed coolant for the lead-cooled fast reactor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only