Gallium Indium alloy, 99.99% (metals basis)
Thermo Scientific Chemicals

Gallium Indium alloy, 99.99% (metals basis)

Ga:In; 40:60 wt.%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 044240.03
also known as 044240-03
Price (USD)
44.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.30
Each
Specifications
Assay Percent Range99.99% (metals basis)
Chemical Name or MaterialGallium Indium alloy
ColorGray
EINECS Number231-163-8
Health Hazard 1H290-H314-H335
View more
Step soldering involving temperature sensitive components, Soldering to molded interconnect device (MID) plastics, Fusible alloys/fuse applications, Mercury replacement, Thermal and electrical conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only