n-Pentane, capillary GC grade, 98+%
n-Pentane, capillary GC grade, 98+%
n-Pentane, capillary GC grade, 98+%
Thermo Scientific Chemicals

n-Pentane, capillary GC grade, 98+%

For organic residue analysis | CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
4 x 4 L
Catalog number 044255.M6
also known as 044255-M6
Price (USD)
449.65
Special offer
Online exclusive
Ends: 28-Dec-2026
529.00
Save 79.35
Each
Quantity:
4 x 4 L
Request bulk or custom format
Price (USD)
449.65
Special offer
Online exclusive
Ends: 28-Dec-2026
529.00
Save 79.35
Each
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Water Content (Karl Fischer Titration)≤0.02%
Residue on Evaporation≤1 ppm
Color (APHA)≤10
Assay (GC)≥98% (as n-Pentane)
View more
For organic residue analysis.n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only