Magnesium iodide, ultra dry, 99.996% (metals basis)
Magnesium iodide, ultra dry, 99.996% (metals basis)
Magnesium iodide, ultra dry, 99.996% (metals basis)
Thermo Scientific Chemicals

Magnesium iodide, ultra dry, 99.996% (metals basis)

Light Sensitive and Hygroscopic | CAS: 10377-58-9 | I2Mg | 278.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 044310.14
also known as 044310-14
Price (USD)
457.65
Online Exclusive
508.00
Save 50.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
457.65
Online Exclusive
508.00
Save 50.35 (10%)
Each
Chemical Identifiers
CAS10377-58-9
IUPAC Namemagnesium(2+) diiodide
Molecular FormulaI2Mg
InChI KeyBLQJIBCZHWBKSL-UHFFFAOYSA-L
SMILES[Mg++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Total Metal Impurities0.004% max.
Residual water< 100 ppm
Magnesium iodide is used to prepare compounds for organic synthesis. It serves as a useful reagent in the Baylis-Hillman reaction. It reacts with cyclic sulfates of vic-diols in order to prepare the corresponding olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only