Nickel(II) perchlorate hydrate, 99.998% (metals basis)
Nickel(II) perchlorate hydrate, 99.998% (metals basis)
Nickel(II) perchlorate hydrate, 99.998% (metals basis)
Thermo Scientific Chemicals

Nickel(II) perchlorate hydrate, 99.998% (metals basis)

CAS: 13637-71-3 | Cl2NiO8 | 257.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044316.18
also known as 044316-18
Price (USD)
224.65
Online Exclusive
249.00
Save 24.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
224.65
Online Exclusive
249.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS13637-71-3
IUPAC Namenickel(2+) diperchlorate
Molecular FormulaCl2NiO8
InChI KeyZLQBNKOPBDZKDP-UHFFFAOYSA-L
SMILES[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Applied as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only