Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)
Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)
Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)

CAS: 13478-33-6 | Cl2CoO8 | 257.825 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044320.09
also known as 044320-09
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Chemical Identifiers
CAS13478-33-6
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only