Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.9999% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.9999% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.9999% (metals basis)
Thermo Scientific Chemicals

Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.9999% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044326.09
also known as 044326-09
Price (USD)
98.65
Special offer
Online exclusive
Ends: 30-Jun-2026
116.00
Save 17.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
98.65
Special offer
Online exclusive
Ends: 30-Jun-2026
116.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metalsless than 0.0001% trace metals
Appearance (Color)Silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only