Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25  in.) length, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25  in.) length, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25  in.) length, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
044327.18
also known as 044327-18
50 g
Catalog number 044327.18
also known as 044327-18
Price (USD)
453.65
Special offer
Online exclusive
Ends: 30-Jun-2026
534.00
Save 80.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
453.65
Special offer
Online exclusive
Ends: 30-Jun-2026
534.00
Save 80.35 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
Assay from Supplier's CofA≥99.9998% (metals basis)
FormSlug, 3.175mm (0.125in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only