Aluminum wire, 0.5mm (0.02 in.) dia., annealed, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 0.5mm (0.02 in.) dia., annealed, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 0.5mm (0.02 in.) dia., annealed, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum wire, 0.5mm (0.02 in.) dia., annealed, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
044333.G6
also known as 044333-G6
10 m
Catalog number 044333.G6
also known as 044333-G6
Price (USD)
382.65
Online Exclusive
425.00
Save 42.35 (10%)
Each
Quantity:
10 m
Request bulk or custom format
Price (USD)
382.65
Online Exclusive
425.00
Save 42.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7429-90-5
ChEBICHEBI:33629
Chemical Name or MaterialAluminum wire
EINECS Number231-072-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only