Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis)
Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis)
Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis)
Thermo Scientific Chemicals

Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis)

CAS: 10450-59-6 | Ce2(SO4)3·8H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 044357.18
also known as 044357-18
Price (USD)
151.65
Special offer
Online exclusive
Ends: 28-Dec-2026
178.00
Save 26.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
151.65
Special offer
Online exclusive
Ends: 28-Dec-2026
178.00
Save 26.35
Each
Chemical Identifiers
CAS10450-59-6
IUPAC Namedicerium(3+) octahydrate trisulfate
Molecular FormulaCe2H16O20S3
InChI KeyPQBKXYUMEMUVIH-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Cerium(III) sulfate octahydrate used in waterproofing, mildew-proofing, and in dyeing and printing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only