5-Vinyluracil, 97%
5-Vinyluracil, 97%
5-Vinyluracil, 97%
Thermo Scientific Chemicals

5-Vinyluracil, 97%

CAS: 37107-81-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 044379.01
also known as 044379-01
Price (USD)
98.65
Online exclusive
109.00
Save 10.35
Each
Quantity:
0.25 g
Request bulk or custom format
Price (USD)
98.65
Online exclusive
109.00
Save 10.35
Each
Specifications
CAS37107-81-6
Chemical Name or Material5-Vinyluracil
Formula Weight138.12
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only