Silicon powder, crystalline, APS ∣<50nm, 98%, Laser synthesized from vapor phase
Silicon powder, crystalline, APS ∣<50nm, 98%, Laser synthesized from vapor phase
Silicon powder, crystalline, APS ∣<50nm, 98%, Laser synthesized from vapor phase
Thermo Scientific Chemicals

Silicon powder, crystalline, APS ∣<50nm, 98%, Laser synthesized from vapor phase

Form: S.A. 70 to 100m2/g | CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 044384.04
also known as 044384-04
Price (USD)
78.65
Online Exclusive
87.70
Save 9.05 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
87.70
Save 9.05 (10%)
Each
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown-yellow
FormPowder
Assay (special)≥ 97.5% Si
Particle Size| <50nm Average particle size
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only