What color are the Silica gel desiccant, indicating, -6+16 mesh granules when they are activated or saturated?
Silica gel desiccant, indicating, -6+16 mesh granules gradually change from a deep blue in the activated condition to a pale pink in the saturated condition.