Gold sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Gold sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)

CAS: 7440-57-5
Have Questions?
Catalog NumberQuantity
044404.KS
also known as 044404-KS
1 Ea.
Catalog number 044404.KS
also known as 044404-KS
Price (USD)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
CAS7440-57-5
Percent Purity99.99%
Quantity1 Ea.
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.