Zinc telluride, 99.99% (metals basis)
Zinc telluride, 99.99% (metals basis)
Zinc telluride, 99.99% (metals basis)
Thermo Scientific Chemicals

Zinc telluride, 99.99% (metals basis)

CAS: 1315-11-3 | TeZn | 192.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044412.14
also known as 044412-14
Price (USD)
279.65
Special offer
Online exclusive
Ends: 28-Dec-2026
329.00
Save 49.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
279.65
Special offer
Online exclusive
Ends: 28-Dec-2026
329.00
Save 49.35
Each
Chemical Identifiers
CAS1315-11-3
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Zinc telluride can be easily doped, and for this reason it is one of the more common semiconducting materials used in optoelectronics. ZnTe is important for development of various semiconductor devices, including blue LEDs, laser diodes, solar cells, and components of microwave generators. It can be used for solar cells, for example, as a back-surface field layer and p-type semiconductor material for a CdTe/ZnTe structure[6] or in PIN diode structures. The material can also be used as a component of ternary semiconductor compounds.Zinc telluride together with lithium niobate is often used for generation of pulsed terahertz radiation in time-domain terahertz spectroscopy and terahertz imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only