Titanium(IV) oxide, catalyst support
Titanium(IV) oxide, catalyst support
Titanium(IV) oxide, catalyst support
Thermo Scientific Chemicals

Titanium(IV) oxide, catalyst support

Anatase, Typical properties = S.A. > 150m2/g, median pore diameter 140Å, total pore volume 0.38cc/g, packing density 0.90kg/L, contains 1% S (as SO4) | CAS: 1317-70-0 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 044429.30
also known as 044429-30
Price (USD)
82.90
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
82.90
Each
Chemical Identifiers
CAS1317-70-0
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPellets, 1/8 inch
DensityPacking Density: 0.70 - 0.85 g/cc
Pore Volume0.43-0.58 ml/g
Surface Area130-180 m2/g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only