Cerium(III) bromide hydrate, 99.998% (metals basis)
Cerium(III) bromide hydrate, 99.998% (metals basis)
Cerium(III) bromide hydrate, 99.998% (metals basis)
Thermo Scientific Chemicals

Cerium(III) bromide hydrate, 99.998% (metals basis)

CAS: 14457-87-5 | Br3Ce | 379.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044432.09
also known as 044432-09
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.50
Save 11.85 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 30-Jun-2026
80.50
Save 11.85 (15%)
Each
Chemical Identifiers
CAS396654-07-2
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only