Aluminum fluoride, anhydrous, 99.99% (metals basis)
Aluminum fluoride, anhydrous, 99.99% (metals basis)
Aluminum fluoride, anhydrous, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum fluoride, anhydrous, 99.99% (metals basis)

CAS: 7784-18-1 | AlF3
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 044435.30
also known as 044435-30
Price (USD)
1,341.65
Online Exclusive
1,490.00
Save 148.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
1,341.65
Online Exclusive
1,490.00
Save 148.35 (10%)
Each
Chemical Identifiers
CAS7784-18-1
IUPAC Namealuminium(3+) trifluoride
Molecular FormulaAlF3
InChI KeyKLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Total Metal Impurities≤0.01% (≤100ppm)
Elemental AnalysisAluminum, Al: Reported (Theory 32.13%)
Appearance (Color)White
Assay (special)≤99.99% (metals basis)
Aluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only