Barium oxide, 99.5% (metals basis excluding Sr), Sr <500ppm
Barium oxide, 99.5% (metals basis excluding Sr), Sr &lt;500ppm
Barium oxide, 99.5% (metals basis excluding Sr), Sr &lt;500ppm
Barium oxide, 99.5% (metals basis excluding Sr), Sr &lt;500ppm
Thermo Scientific Chemicals

Barium oxide, 99.5% (metals basis excluding Sr), Sr <500ppm

CAS: 1304-28-5 | BaO | 153.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 044440.14
also known as 044440-14
Price (USD)
269.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
269.00
Each
Add to cart
Chemical Identifiers
CAS1304-28-5
IUPAC Nameoxobarium
Molecular FormulaBaO
InChI KeyQVQLCTNNEUAWMS-UHFFFAOYSA-N
SMILESO=[Ba]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSr<500ppm
Total Metal Impurities0.5% max.
Barium oxide is used in cathode ray tubes, crown glass, and catalysts. It is used for the adsorption NO and NO2. It also plays an important role for NO2 storage and reduction. It acts as an efficient ethoxylation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only