Cerium(III) bromide hydrate, 99%
Cerium(III) bromide hydrate, 99%
Cerium(III) bromide hydrate, 99%
Thermo Scientific Chemicals

Cerium(III) bromide hydrate, 99%

CAS: 14457-87-5 | Br3Ce | 379.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 044447.14
also known as 044447-14
Price (USD)
177.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
177.00
Each
Chemical Identifiers
CAS14457-87-5
IUPAC Namecerium(3+) tribromide
Molecular FormulaBr3Ce
InChI KeyMOOUSOJAOQPDEH-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Cerium(III) bromide hydrate is used in the security, medical imaging, and geophysics detectors, γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only