Potassium hexafluorosilicate, 99.999% (metals basis)
Potassium hexafluorosilicate, 99.999% (metals basis)
Potassium hexafluorosilicate, 99.999% (metals basis)
Thermo Scientific Chemicals

Potassium hexafluorosilicate, 99.999% (metals basis)

CAS: 16871-90-2 | F6K2Si | 220.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 044495.09
also known as 044495-09
Price (USD)
66.65
Special offer
Online exclusive
Ends: 28-Dec-2026
78.80
Save 12.15
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 28-Dec-2026
78.80
Save 12.15
Each
Chemical Identifiers
CAS16871-90-2
IUPAC Namedipotassium hexafluorosilanediuide
Molecular FormulaF6K2Si
InChI KeyUUHPPUWEDCVPCO-UHFFFAOYSA-N
SMILES[K+].[K+].F[Si--](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Potassium hexafluorosilicate is used in the production of porceleins. It is also used for the preservation of timber. It is involved in the manufacturing of ceramics, aluminum and magnesium smelting. In addition, it is used as an intermediate in the manufacturing of optical glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only