Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)
Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)
Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)
Thermo Scientific Chemicals

Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)

CAS: 54451-25-1 | C3Ce2O9 | 460.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044496.14
also known as 044496-14
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS54451-25-1
IUPAC Namedicerium(3+) tricarbonate
Molecular FormulaC3Ce2O9
InChI KeyGHLITDDQOMIBFS-UHFFFAOYSA-H
SMILES[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.001% max.
Cerium(III) carbonate hydrate is used to prepare cerium chloride and incandescent lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only