Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
044498.22
also known as 044498-22
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044498.22
also known as 044498-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS681435-25-6
IUPAC Namecadmium(2+) dibromide
Molecular FormulaBr2Cd
InChI KeyKPWJBEFBFLRCLH-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only