2-Hydroxymethyl-15-crown-5, 98%
2-Hydroxymethyl-15-crown-5, 98%
2-Hydroxymethyl-15-crown-5, 98%
Thermo Scientific Chemicals

2-Hydroxymethyl-15-crown-5, 98%

CAS: 75507-25-4 | C11H22O6 | 250.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044500.03
also known as 044500-03
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Specifications
Beilstein3589706
CAS75507-25-4
Chemical Name or Material2-Hydroxymethyl-15-crown-5
Density1.175 g/mL
EINECS Number278-222-4
View more
2-Hydroxymethyl-15-crown-5 is used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only