Amidosulfonic acid, 99.99% (metals basis)
Amidosulfonic acid, 99.99% (metals basis)
Amidosulfonic acid, 99.99% (metals basis)
Thermo Scientific Chemicals

Amidosulfonic acid, 99.99% (metals basis)

CAS: 5329-14-6 | H3NO3S | 97.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044510.14
also known as 044510-14
Price (USD)
29.50
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.50
Each
Chemical Identifiers
CAS5329-14-6
IUPAC Namesulfamic acid
Molecular FormulaH3NO3S
InChI KeyIIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILESNS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Total Metal Impurities≤0.01%
Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only