n-Hexadecane, 99.5+%
n-Hexadecane, 99.5+%
n-Hexadecane, 99.5+%
Thermo Scientific Chemicals

n-Hexadecane, 99.5+%

CAS: 544-76-3 | C16H34 | 226.448 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044511.14
also known as 044511-14
Price (USD)
223.65
Special offer
Online exclusive
Ends: 30-Jun-2026
263.00
Save 39.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
223.65
Special offer
Online exclusive
Ends: 30-Jun-2026
263.00
Save 39.35 (15%)
Each
Chemical Identifiers
CAS544-76-3
IUPAC Namehexadecane
Molecular FormulaC16H34
InChI KeyDCAYPVUWAIABOU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>/= 99.5%
Appearance (Color)Clear, colorless
FormLiquid
Employed in industry as Adsorbents, Fuels and fuel additives. Also in Fabric, Textile, and Leather Products. Solvent, organic intermediate, ignition standard for diesel fuels. Fuels and Related Products. Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only